1-Bromo-3-chloro-5-fluoro-4-nitrobenzene 95%

Supplier: Ambeed
Danger

A309119-250MG A309119-25G A309119-5G A309119-1G
77114-568EA 28.65 USD
77114-568 77593-848 77593-850 77685-883
1-Bromo-3-chloro-5-fluoro-4-nitrobenzene 95%
1-Bromo-3-chloro-5-fluoro-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClFNO₂
MW: 254.44 g/mol
MDL Number: MFCD27947500
CAS Number: 218797-71-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR