(2S,4S)-4-Aminopyrrolidine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: H-cis-4-NH₂-Pro-OH

A308091-1G A308091-250MG
77114-554EA 434.97 USD
77114-554 77114-556
(2S,4S)-4-Aminopyrrolidine-2-carboxylic acid 95%
(2S,4S)-4-Aminopyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₁₀N₂O₂
MW: 130.15 g/mol
MDL Number: MFCD08704537
CAS Number: 16257-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR