4-Amino-N-methylindole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-indol-4-amine

A306115-1G A306115-250MG
77114-534EA 30.91 USD
77114-534 77114-536
4-Amino-N-methylindole 98%
4-Amino-N-methylindole

Precaución: for Research Use Only

Formula: C₉H₁₀N₂
MDL Number: MFCD08061153
CAS Number: 85696-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR