3-Ethyl-4-methyl-3-pyrrolin-2-one 98%

Supplier: Ambeed
Warning

Synonyms: 3-Ethyl-4-methyl-2,5-dihydro-1H-pyrrol-2-one, 1,5-Dihydro-3-ethyl-4-methyl-2H-pyrrol-2-one

A304885-100G A304885-10G A304885-25G A304885-5G
77114-518EA 68.96 USD
77114-518 77114-520 77114-522 77114-524
3-Ethyl-4-methyl-3-pyrrolin-2-one 98%
3-Ethyl-4-methyl-3-pyrrolin-2-one

Precaución: for Research Use Only

Formula: C₇H₁₁NO
MW: 125.17 g/mol
MDL Number: MFCD00173861
CAS Number: 766-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR