α,2,4-Trichlorotoluene 98%

Supplier: Ambeed
Danger

A303672-10G A303672-25G A303672-500G
77114-510EA 16.46 USD
77114-510 77114-512 77114-514
α,2,4-Trichlorotoluene 98%
α,2,4-Trichlorotoluene

Precaución: for Research Use Only

Formula: C₇H₅Cl₃
MDL Number: MFCD00000895
CAS Number: 94-99-5
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR