N1,N2-Diphenethyloxalamide 95%

Supplier: Ambeed
Danger

A302115-100G A302115-1G A302115-250MG A302115-25G A302115-5G
77114-488EA 2497.36 USD
77114-488 77114-490 77114-492 77114-494 77114-496
N1,N2-Diphenethyloxalamide 95%
N1,N2-Diphenethyloxalamide

Precaución: for Research Use Only

Formula: C₁₈H₂₀N₂O₂
MW: 296.36 g/mol
MDL Number: MFCD00030029
CAS Number: 14040-79-0
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR