TC-H 106 98%

Supplier: Ambeed
Warning

Synonyms: N1-(2-Aminophenyl)-N7-(p-tolyl)heptanediamide, Pimelic Diphenylamide 106, N1-(2-Aminophenyl)-N7-(4-methylphenyl)heptanediamide

A301260-100MG A301260-10MG A301260-1MG A301260-25MG A301260-50MG A301260-5MG
77114-470EA 82.4 USD
77114-470 77114-472 77114-474 77114-476 77114-478 77114-480
TC-H 106 98%
TC-H 106

Precaución: for Research Use Only

Formula: C₂₀H₂₅N₃O₂
MW: 339.43 g/mol
MDL Number: MFCD17010287
CAS Number: 937039-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR