Methyl-2-(2-amino-1,3-thiazol-4-yl)acetate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-(2-aminothiazol-4-yl)acetate
A300401-10G
A300401-1G
A300401-250MG
A300401-25G
A300401-5G
77114-456EA
99.73
USD
77114-456
77114-458
77114-460
77114-462
77114-464
Methyl-2-(2-amino-1,3-thiazol-4-yl)acetate 95%
Methyl-2-(2-amino-1,3-thiazol-4-yl)acetate
Precaución: for Research Use Only
Formula:
C₆H₈N₂O₂S MW: 172.21 g/mol |
MDL Number:
MFCD00129969 CAS Number: 64987-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...