Diacetone acrylamide 98% stabilized
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(2-Methyl-4-oxopentan-2-yl)acrylamide, N-(1,1-Dimethyl-3-oxobutyl)acrylamide
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
A298990-1000G
A298990-100G
A298990-25G
A298990-500G
77114-434EA
62.6
USD
77114-434
77114-436
77114-438
77114-440
Diacetone acrylamide 98% stabilized
Diacetone acrylamide
Precaución: for Research Use Only
Formula:
C₉H₁₅NO₂ MW: 169.22 g/mol |
MDL Number:
MFCD00008788 CAS Number: 2873-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...