3-Fluoro-2-methoxypyridine 97%

Supplier: Ambeed
Danger

A298858-10G A298858-1G A298858-5G
77114-428EA 45.32 USD
77114-428 77114-430 77114-432
3-Fluoro-2-methoxypyridine 97%
3-Fluoro-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₆FNO
MDL Number: MFCD03094945
CAS Number: 884494-69-3
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR