Methyl-2,4-difluorobenzoate 98%

Supplier: Ambeed
Warning

A295155-100G A295155-10G A295155-25G A295155-5G
77114-356EA 46.35 USD
77114-356 77114-358 77114-360 77114-362
Methyl-2,4-difluorobenzoate 98%
Methyl-2,4-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MDL Number: MFCD03093799
CAS Number: 106614-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR