1,2-Dimethyl-3-fluoro-4-iodobenzene 95%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-1-iodo-3,4-dimethylbenzene

A295042-100MG A295042-250MG
77114-352EA 59.42 USD
77114-352 77114-354
1,2-Dimethyl-3-fluoro-4-iodobenzene 95%
1,2-Dimethyl-3-fluoro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₈FI
MW: 250.05 g/mol
MDL Number: MFCD28794424
CAS Number: 1806323-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR