2,6-Dichloro-3-nitrobenzaldehyde 98%

Supplier: Ambeed
Danger

A294623-100G A294623-1G A294623-25G A294623-5G
77114-344EA 216.3 USD
77114-344 77114-346 77114-348 77114-350
2,6-Dichloro-3-nitrobenzaldehyde 98%
2,6-Dichloro-3-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Cl₂NO₃
MDL Number: MFCD00090332
CAS Number: 5866-97-7
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR