Isonicotinic acid-N-oxide 98%

Supplier: Ambeed
Warning

Synonyms: Isonicotinic acid N-oxide, 1-Oxidopyridin-1-ium-4-carboxylic acid, 4-Carboxypyridine 1-oxide, Pyridine-4-carboxylic acid N-oxide

A294315-100G A294315-25G A294315-500G A294315-5G
77114-328EA 77.45 USD
77114-328 77114-330 77114-332 77114-334
Isonicotinic acid-N-oxide 98%
Isonicotinic acid N-oxide

Precaución: for Research Use Only

Formula: C₆H₅NO₃
MW: 139.11 g/mol
MDL Number: MFCD00006209
CAS Number: 13602-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR