3-(Aminomethyl)phenylboronic acid hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (3-(Aminomethyl)phenyl)boronic acid hydrochloride, 3-(Aminomethyl)benzeneboronic acid hydrochloride

A294096-1G A294096-250MG A294096-5G
77114-322EA 38.12 USD
77114-322 77114-324 77114-326
3-(Aminomethyl)phenylboronic acid hydrochloride 95%
3-(Aminomethyl)phenylboronic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₁BClNO₂
MW: 187.43 g/mol
MDL Number: MFCD02093050
CAS Number: 352525-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR