Sulfadiazine 98%

Supplier: Ambeed
Danger

Synonyms: N-(2-Pyrimidinyl)sulphanilamide, 4-Amino-N-(pyrimidin-2-yl)benzenesulphonamide, 4-Amino-N-(2-pyrimidinyl)benzenesulphonamide

A293414-1000G A293414-100G A293414-25G A293414-500G
77114-310EA 166.86 USD
77114-310 77114-312 77114-314 77612-690
Sulfadiazine 98%
Sulfadiazine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₄O₂S
MW: 250.28 g/mol
MDL Number: MFCD00006065
CAS Number: 68-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR