3-Iodo-p-anisidine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Iodo-4-methoxyaniline

A293237-10G A293237-1G A293237-25G A293237-5G
77114-304EA 63.66 USD
77114-304 77114-306 77114-308 77685-835
3-Iodo-p-anisidine 98%
3-Iodo-p-anisidine

Precaución: for Research Use Only

Formula: C₇H₈INO
MW: 249.0490 g/mol
MDL Number: MFCD06656566
CAS Number: 74587-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR