Methyl 5-amino-4-fluoro-1-methyl-1H-1,3-benzodiazole-6-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-amino-4-fluoro-1-methyl-1H-benzo[d]imidazole-6-carboxylate

A292179-100MG A292179-1G A292179-250MG A292179-5G
77114-270EA 8.49 USD
77114-270 77114-272 77114-274 77114-276
Methyl 5-amino-4-fluoro-1-methyl-1H-1,3-benzodiazole-6-carboxylate 97%
Methyl 5-amino-4-fluoro-1-methyl-1H-1,3-benzodiazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀FN₃O₂
MW: 223.204 g/mol
MDL Number: MFCD26127252
CAS Number: 918321-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR