3-Amino-2-chloro-6-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-2-chloro-6-picoline, 5-Amino-6-chloro-2-picoline

A291715-1G A291715-250MG A291715-5G A291715-10G A291715-25G A291715-100G
77114-264EA 15.45 USD
77114-264 77114-266 77114-268 77685-832 77685-833 77685-831
3-Amino-2-chloro-6-methylpyridine 98%
3-Amino-2-chloro-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MW: 142.5900 g/mol
MDL Number: MFCD03095213
CAS Number: 39745-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR