1-(Chloro(piperidin-1-yl)methylene)piperidin-1-ium hexafluorophosphate(V) 98%

Supplier: Ambeed
Warning

A290226-1G A290226-5G
77114-252EA 20.16 USD
77114-252 77114-254
1-(Chloro(piperidin-1-yl)methylene)piperidin-1-ium hexafluorophosphate(V) 98%
1-(Chloro(piperidin-1-yl)methylene)piperidin-1-ium hexafluorophosphate(V)

Precaución: for Research Use Only

Formula: C₁₁H₂₀ClF₆N₂P
MW: 360.70 g/mol
MDL Number: MFCD00191817
CAS Number: 161308-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR