Boc-D-allo-Isoleucine 98%

Supplier: Ambeed
Warning

Synonyms: Boc-D-allo-Ile-OH, (2R,3S)-2-((tert-Butoxycarbonyl)amino)-3-methylpentanoic acid

A289231-100MG A289231-250MG A289231-5G A289231-1G
77114-244EA 14.42 USD
77114-244 77114-246 77114-248 77685-828
Boc-D-allo-Isoleucine 98%
Boc-D-allo-Isoleucine

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₄
MW: 231.292 g/mol
MDL Number: MFCD00237543
CAS Number: 55780-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR