2-Amino-5-methylthiophene-3-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-methyl-3-thiophenecarbonitrile, 2-Amino-3-cyano-5-methylthiophene

A289215-100G A289215-10G A289215-1G A289215-25G A289215-5G
77114-234EA 125.19 USD
77114-234 77114-236 77114-238 77114-240 77114-242
2-Amino-5-methylthiophene-3-carbonitrile 97%
2-Amino-5-methylthiophene-3-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₆N₂S
MW: 138.1902 g/mol
MDL Number: MFCD01626055
CAS Number: 138564-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR