Isoliquiritigenin 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
(E)-1-(2,4-Dihydroxyphenyl)-3-(4-hydroxyphenyl)prop-2-en-1-one, 2',4,4'-Trihydroxychalcone, (E)-1-(2,4-Dihydroxyphenyl)-3-(4-hydroxyphenyl)-2-propen-1-one
A289179-100MG
A289179-10MG
A289179-1G
A289179-250MG
A289179-50MG
A289179-1MG
A289179-5G
77114-224EA
18.04
USD
77114-224
77114-226
77114-228
77114-230
77114-232
77685-826
77685-827
Isoliquiritigenin 98+%
Isoliquiritigenin
Precaución: for Research Use Only
Formula:
C₁₅H₁₂O₄ MW: 256.26 g/mol |
MDL Number:
MFCD00075907 CAS Number: 961-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...