2-Bromoethyl acetate 98%

Supplier: Ambeed
Warning

Synonyms: Acetic acid (2-bromoethyl) ester

A288884-1000G A288884-100G A288884-10G A288884-1G A288884-500G A288884-5G A288884-25G
77114-212EA 219.61 USD
77114-212 77114-214 77114-216 77114-218 77114-220 77114-222 77305-546
2-Bromoethyl acetate 98%
2-Bromoethyl acetate

Precaución: for Research Use Only

Formula: C₄H₇BrO₂
MW: 167 g/mol
MDL Number: MFCD00000235
CAS Number: 927-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR