Miltefosine 98%

Supplier: Ambeed
Danger

Synonyms: Hexadecyl 2-(Trimethylammonio)ethyl phosphate, Phosphoric acid hexadecyl 2-(trimethylammonio)ethyl ester, Fos-Choline™ 16, HePC, N-Hexadecylphosphocholine, Hexadecyl Phosphorylcholine

A288826-1G A288826-25MG A288826-5G A288826-250MG A288826-25G A288826-1MG
77114-206EA 43.5 USD
77114-206 77114-208 77114-210 77616-244 77616-246 77685-824
Miltefosine 98%
Miltefosine

Precaución: for Research Use Only

Formula: C₂₁H₄₆NO₄P
MW: 407.57 g/mol
MDL Number: MFCD00133396
CAS Number: 58066-85-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR