SAR-131675 95+%
Supplier: Ambeed
|
Warning
|
Synonyms:
(-)-2-Amino-1-ethyl-7-(3-hydroxy-4-methoxy-3-methylbut-1-yn-1-yl)-N-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxamide
A286832-10MG
A286832-1MG
A286832-25MG
77114-180EA
58.35
USD
77114-180
77114-182
77114-184
SAR-131675 95+%
SAR-131675
Precaución: for Research Use Only
Formula:
C₁₈H₂₂N₄O₄ MW: 358.4 g/mol |
MDL Number:
MFCD23098773 CAS Number: 1092539-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...