4-Bromo-2-nitro-6-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-bromo-3-nitrobenzotrifluoride

A285694-1G A285694-5G
77114-152EA 14.42 USD
77114-152 77114-154
4-Bromo-2-nitro-6-(trifluoromethyl)aniline 98%
4-Bromo-2-nitro-6-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄BrF₃N₂O₂
MW: 285.02 g/mol
MDL Number: MFCD09835318
CAS Number: 157026-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR