3-Bromophenylhydrazinium chloride 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromophenylhydrazine hydrochloride, 1-(3-Bromophenyl)hydrazine hydrochloride

A285676-100G A285676-10G A285676-25G A285676-500G A285676-5G
77114-142EA 100.94 USD
77114-142 77114-144 77114-146 77114-148 77114-150
3-Bromophenylhydrazinium chloride 98%
3-Bromophenylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₆H₈BrClN₂
MW: 223.5
MDL Number: MFCD00012933
CAS Number: 27246-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR