2,3-Dichlorobenzonitrile 98%

Supplier: Ambeed
Warning

A285610-100G A285610-1G A285610-25G A285610-5G A285610-10G A285610-500G
77114-134EA 77.25 USD
77114-134 77114-136 77114-138 77114-140 77685-812 77685-813
2,3-Dichlorobenzonitrile 98%
2,3-Dichlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃Cl₂N
MDL Number: MFCD00016372
CAS Number: 6574-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR