4-Oxo-2-spiro-(N-boc-piperidine-4-yl)benzopyran 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-oxospiro[chroman-2,4'-piperidine]-1'-carboxylate

A284737-1G A284737-250MG A284737-5G
77114-124EA 49.44 USD
77114-124 77114-126 77114-128
4-Oxo-2-spiro-(N-boc-piperidine-4-yl)benzopyran 98%
4-Oxo-2-spiro-(N-boc-piperidine-4-yl)benzopyran

Precaución: for Research Use Only

Formula: C₁₈H₂₃NO₄
MDL Number: MFCD05861626
CAS Number: 849928-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR