2-(5-Nitro-1H-imidazol-1-yl)ethanol 95%

Supplier: Ambeed
Warning

A283247-100MG A283247-250MG
77114-104EA 135.96 USD
77114-104 77114-106
2-(5-Nitro-1H-imidazol-1-yl)ethanol 95%
2-(5-Nitro-1H-imidazol-1-yl)ethanol

Precaución: for Research Use Only

Formula: C₅H₇N₃O₃
MDL Number: MFCD00159670
CAS Number: 5006-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR