1-Cbz-4-methylpiperidine-4-carboxylic acid 98%

Supplier: Ambeed
Danger

A282904-1G A282904-250MG A282904-5G A282904-25G
77114-098EA 43.26 USD
77114-098 77114-100 77114-102 77571-752
1-Cbz-4-methylpiperidine-4-carboxylic acid 98%
1-Cbz-4-methylpiperidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₄
MDL Number: MFCD11100934
CAS Number: 203522-12-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR