5-Bromo-2-fluoroanisole 97%

Supplier: Ambeed
Warning

A279039-10G A279039-25G A279039-500G A279039-5G A279039-100G
77114-024EA 19.57 USD
77114-024 77114-026 77114-028 77114-030 77616-180
5-Bromo-2-fluoroanisole 97%
5-Bromo-2-fluoroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrFO
MDL Number: MFCD01861130
CAS Number: 103291-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR