8-Hydroxy-1,2,3,4-tetrahydroquinoline 95%

Supplier: Ambeed
Warning

Synonyms: 1,2,3,4-Tetrahydroquinolin-8-ol, 1,2,3,4-Tetrahydro-8-hydroxyquinoline

A278198-10G A278198-1G A278198-250MG A278198-25G A278198-5G
77114-014EA 142.17 USD
77114-014 77114-016 77114-018 77114-020 77114-022
8-Hydroxy-1,2,3,4-tetrahydroquinoline 95%
8-Hydroxy-1,2,3,4-tetrahydroquinoline

Precaución: for Research Use Only

Formula: C₉H₁₁NO
MW: 149.1900 g/mol
MDL Number: MFCD02656029
CAS Number: 6640-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR