5,7-Dimethyl-8-quinolinol 97%

Supplier: Ambeed
Warning

Synonyms: 5,7-Dimethyl-8-hydroxyquinoline

A278090-100MG A278090-1G A278090-250MG A278090-5G
77114-006EA 92.3 USD
77114-006 77114-008 77114-010 77114-012
5,7-Dimethyl-8-quinolinol 97%
5,7-Dimethyl-8-quinolinol

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO
MW: 173.2111 g/mol
MDL Number: MFCD00168976
CAS Number: 37873-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR