4-Fluoro-3-nitrobenzotrifluoride 98%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-3-Nitrobenzotrifluoride

A277073-100G A277073-10G A277073-25G A277073-500G A277073-5G
77113-968EA 27.81 USD
77113-968 77113-970 77113-972 77113-974 77113-976
4-Fluoro-3-nitrobenzotrifluoride 98%
4-Fluoro-3-nitrobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃F₄NO₂
MDL Number: MFCD00007059
CAS Number: 367-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR