4-(Aminomethyl)benzenesulfonamide hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: alpha-Amino-p-toluenesulphonamide hydrochloride, Homosulphamine hydrochloride, Mafenide hydrochloride, Mafenide HCl

A275314-100G A275314-1G A275314-5G
77113-956EA 147.29 USD
77113-956 77113-958 77113-960 77571-698 77685-778
4-(Aminomethyl)benzenesulfonamide hydrochloride 98%
4-(Aminomethyl)benzenesulfonamide hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₁ClN₂O₂S
MW: 222.6924 g/mol
MDL Number: MFCD00013005
CAS Number: 138-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR