1-Bromo-4-chloro-2-(difluoromethoxy)benzene 95%
Supplier: Ambeed
|
Warning
|
A274545-100MG
A274545-250MG
77113-952EA
108.22
USD
77113-952
77113-954
1-Bromo-4-chloro-2-(difluoromethoxy)benzene 95%
1-Bromo-4-chloro-2-(difluoromethoxy)benzene
Precaución: for Research Use Only
Formula:
C₇H₄BrClF₂O MW: 257.45 g/mol |
MDL Number:
MFCD14698391 CAS Number: 1056942-37-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...