2,3,5-Trichlorobenzoic acid 95%

Supplier: Ambeed
Warning

A273977-100G A273977-10G A273977-25G A273977-5G
77113-930EA 1118.19 USD
77113-930 77113-932 77113-934 77113-936
2,3,5-Trichlorobenzoic acid 95%
2,3,5-Trichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Cl₃O₂
MW: 225.46 g/mol
MDL Number: MFCD00060674
CAS Number: 50-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR