1,3-Bis(3,4-dichlorophenyl)urea 95%

Supplier: Ambeed
Warning

Synonyms: 3,3',4,4'-Tetrachlorocarbanilide

A273349-1G A273349-250MG A273349-5G
77113-924EA 61.54 USD
77113-924 77113-926 77113-928
1,3-Bis(3,4-dichlorophenyl)urea 95%
1,3-Bis(3,4-dichlorophenyl)urea

Precaución: for Research Use Only

Formula: C₁₃H₈Cl₄N₂O
MW: 350.027 g/mol
MDL Number: MFCD00171464
CAS Number: 4300-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR