2-Bromo-5-fluoro-3-nitropyridine 98%

Supplier: Ambeed
Warning

A272553-10G A272553-1G
77113-918EA 142.17 USD
77113-918 77685-772
2-Bromo-5-fluoro-3-nitropyridine 98%
2-Bromo-5-fluoro-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrFN₂O₂
MW: 220.99 g/mol
MDL Number: MFCD05662387
CAS Number: 652160-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR