Oxalic acid dihydrazide 98%

Supplier: Ambeed
Danger

Synonyms: Oxalohydrazide, Oxalyl dihydrazide, Oxalic dihydrazide, Ethanedioic acid dihydrazide

A270444-25G A270444-5G
77113-896EA 15.92 USD
77113-896 77113-898
Oxalic acid dihydrazide 98%
Oxalic acid dihydrazide

Precaución: for Research Use Only

Formula: C₂H₆N₄O₂
MW: 118.1 g/mol
MDL Number: MFCD00007608
CAS Number: 996-98-5
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR