SU 9516 98+%

Supplier: Ambeed
Warning

Synonyms: SU9516, (Z)-3-((1H-Imidazol-4-yl)methylene)-5-methoxyindolin-2-one, 3-[1-(3H-Imidazol-4-yl)-meth-(Z)-ylidene]-5-methoxy-1,3-dihydro-indol-2-one

A269439-10MG A269439-1MG A269439-25MG A269439-50MG A269439-5MG
77113-872EA 42.44 USD
77113-872 77113-874 77113-876 77113-878 77113-880
SU 9516 98+%
SU 9516

Precaución: for Research Use Only

Formula: C₁₃H₁₁N₃O₂
MW: 241.3 g/mol
MDL Number: MFCD17010284
CAS Number: 377090-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR