(R)-2-Aminobut-3-en-1-ol hydrochloride 97%

Supplier: Ambeed
Warning

A268476-100MG A268476-1G A268476-250MG
77113-862EA 35.02 USD
77113-862 77113-864 77113-866
(R)-2-Aminobut-3-en-1-ol hydrochloride 97%
(R)-2-Aminobut-3-en-1-ol hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClNO
MDL Number: MFCD09025317
CAS Number: 313995-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR