3-Amino-2-cyclohexen-1-one 98%

Supplier: Ambeed
Warning

A267487-100G A267487-10G A267487-25G A267487-5G
77113-830EA 90.64 USD
77113-830 77113-832 77113-834 77113-836
3-Amino-2-cyclohexen-1-one 98%
3-Amino-2-cyclohexen-1-one

Precaución: for Research Use Only

Formula: C₆H₉NO
MDL Number: MFCD00013783
CAS Number: 5220-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR