Nicotinic acid hydrazide 98%

Supplier: Ambeed
Warning

Synonyms: Nicotinohydrazide, (3-Pyridylcarbonyl)hydrazine, Pyridine-3-carbohydrazide

A266061-100G A266061-10G A266061-25G A266061-5G
77113-810EA 45.32 USD
77113-810 77113-812 77113-814 77685-760
Nicotinic acid hydrazide 98%
Nicotinic acid hydrazide

Precaución: for Research Use Only

Formula: C₆H₇N₃O
MDL Number: MFCD00006383
CAS Number: 553-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR