2-Chloro-3,5-dibromo-4-methylpyridine 95%

Supplier: Ambeed
Warning

A264354-1G A264354-250MG A264354-5G
77113-758EA 55.17 USD
77113-758 77113-760 77113-762
2-Chloro-3,5-dibromo-4-methylpyridine 95%
2-Chloro-3,5-dibromo-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₄Br₂ClN
MW: 285.36 g/mol
MDL Number: MFCD09864796
CAS Number: 1000017-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR