1-(3-aminopropyl)pyrrolidin-2-one 95%

Supplier: Ambeed
Danger

Synonyms: N-(3'-Aminopropyl)-2-pyrrolidinone, 1-(3-Aminopropyl)pyrrolidin-2-one

A263936-1G A263936-25G A263936-5G
77113-730EA 30.77 USD
77113-730 77113-732 77113-734
1-(3-aminopropyl)pyrrolidin-2-one 95%
1-(3-aminopropyl)pyrrolidin-2-one

Precaución: for Research Use Only

Formula: C₇H₁₄N₂O
MW: 142.202 g/mol
MDL Number: MFCD00003201
CAS Number: 7663-77-6
UN: 3267
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR