1-Bromo-3-(difluoromethoxy)benzene 98%

Supplier: Ambeed
Warning

A263176-100G A263176-10G A263176-1G A263176-25G A263176-5G
77113-720EA 214.31 USD
77113-720 77113-722 77113-724 77113-726 77113-728
1-Bromo-3-(difluoromethoxy)benzene 98%
1-Bromo-3-(difluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂O
MW: 223.02 g/mol
MDL Number: MFCD00236233
CAS Number: 262587-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR