Ethyl-4-amino-3-chlorobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-amino-3-chlorobenzoate, 4-Amino-3-chlorobenzoic acid ethyl ester

A262604-1G A262604-250MG A262604-5G
77113-712EA 21.22 USD
77113-712 77113-714 77113-716
Ethyl-4-amino-3-chlorobenzoate 97%
Ethyl 4-amino-3-chlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.6300 g/mol
MDL Number: MFCD06411007
CAS Number: 82765-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR